Calcium nitrate tetrahydrate 98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC44753-0250 AC44753-0010 44753-0050
TS44753-0250EA 549 USD
TS44753-0250 TS44753-0010 TS44753-0050
Calcium nitrate tetrahydrate 98%, Extra Pure
Calcium nitrate tetrahydrate
Formula: Ca(NO₃)₂·4H₂O
MW: 236.15 g/mol
Boiling Pt: 132 °C (760 mmHg)
Melting Pt: 45 °C
Density: 1.86 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 13477-34-4

Order Now

Specification Test Results

Appearance White Crystals
Titration Complexometric ≥97.5 %
pH 5.0 to 9.0
Chloride (Cl) ≤0.01 %
Sulfate (SO4) ≤0.01 %
Iron (Fe) ≤0.002 %
Heavy metals (as Pb) ≤0.002 %
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR