Disodium hydrogen phosphate heptahydrate 98+% ACS
Supplier: Thermo Scientific Chemicals
42438-0010
AC42438-0050
AC42438-5000
TS42438-0010EA
162
USD
TS42438-0010
TS42438-0050
TS42438-5000
Disodium hydrogen phosphate heptahydrate 98+% ACS
di-Sodium hydrogen phosphate heptahydrate
Formula:
HNa₂O₄P·7H₂O Melting Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149180 CAS Number: 7782-85-6 EINECS: 231-448-7 |
Specification Test Results
Appearance | Colorless to white Granular powder or crystals |
pH | 8.7 to 9.3 (5 % solution at 25.0°C) |
Titration excess HCl | 98.0 to 102.0 % |
Chloride (Cl) | ≤10 ppm |
Sulfate (SO4) | ≤50 ppm |
Iron (Fe) | ≤10 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...