Disodium hydrogen phosphate heptahydrate ca. 95% for analysis

Supplier: Thermo Scientific Chemicals
Danger

20651-5000 20651-0025 20651-2500
TS20651-5000EA 111 USD
TS20651-5000 TS20651-0025 TS20651-2500
Disodium hydrogen phosphate heptahydrate ca. 95% for analysis
di-Sodium hydrogen phosphate heptahydrate
Formula: HNa₂O₄P·7H₂O
Boiling Pt: 169…171 °C
Melting Pt: –66 °C
Density: 0.741 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS Number: 7782-85-6
EINECS: 212-819-2
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4220 (20°C, 589 nm)
GC ≥94 %
Titration with HClO4 ≥97.5 %
Arsenic (As) ≤5 ppm
Iron (Fe) ≤5 ppm
Potassium (K) ≤100 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR