Disodium hydrogen phosphate dihydrate 99+% for HPLC

Supplier: Thermo Scientific Chemicals

34381-0025 34381-5000 AC34381-0250 AC34381-1000
TS34381-0025EA 743 USD
TS34381-0025 TS34381-5000 TS34381-0250 TS34381-1000
Disodium hydrogen phosphate dihydrate 99+% for HPLC
di-Sodium hydrogen phosphate dihydrate
Formula: Na₂HPO₄·2H₂O
Melting Pt: 243…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00149182
CAS Number: 10028-24-7
EINECS: 231-448-7

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
UV at 254 nm A: ≤0.05
pH 7.5 to 9.2 (5 % at 25°C)
Titration with HCl ≥99.0 %
Chloride (Cl) ≤50 ppm
Lead (Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR