Tetrahydrofuran-[D₈] 99.5 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: THF-d8THF-D8OctadeuterotetrahydrofuranTetrahydrofuran-d8Deuterated TetrahydrofuranTetrahydrofuran-[²H₈]

32078-0075
TS32078-0075EA 388 USD
TS32078-0075
Tetrahydrofuran-[D₈] 99.5 atom % D for NMR spectroscopy
Tetrahydrofuran-[D8]
Formula: C₄D₈O
MW: 80.04 g/mol
Boiling Pt: 65 °C (1013 hPa)
Melting Pt: –106 °C
Density: 0.98 g/cm³
Flash Pt: –21 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00044238
CAS Number: 1693-74-9
EINECS: 216-898-4
UN: 2056
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 % (Coulometric)
D-Enrichment ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR