Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dichloromethane-[²H₂]
, Dideuteromethylene chloride
, Methylene chloride-D2
, Methylene chloride-d2
, Dichloromethane-d2
32072-0075
TS32072-0075EA
275.97
USD
TS32072-0075
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Dichloromethane-[D2]
Formula:
CD₂Cl₂ MW: 86.92 g/mol Boiling Pt: 40 °C (1013 hPa) Melting Pt: –97 °C Density: 1.36 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000882 CAS Number: 1665-00-5 EINECS: 216-776-0 UN: 1593 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.02 % (Coulometric) |
D-Enrichment | ≥99.75 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...