Potassium 98%, chunks in mineral oil
Supplier: Thermo Scientific Chemicals
|
Danger
|
223865000
223860100
22386-1000
22386-0500
TS22386-0500EA
177.3
USD
TS22386-0500
TS223865000
TS223860100
TS22386-1000
Potassium 98%, chunks in mineral oil
Potassium
Formula:
K MW: 39.1 g/mol Boiling Pt: 770 °C (760 mmHg) Melting Pt: 63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00133776 CAS Number: 7440-09-7 EINECS: 231-119-8 UN: 2257 ADR: 4.3,I |
Specification Test Results
Appearance | Gray Chunks |
Total trace metal impurities | ≤2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...