Aluminium oxide 99.7+%, Extra Pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Aluminium(III) oxide
, Alumina C
AC21570-0050
AC21570-0010
AC21570-2500
TS21570-2500EA
28.8
USD
TS21570-2500
TS21570-0010
TS21570-0050
Aluminium oxide 99.7+%, Extra Pure
Aluminium oxide
Formula:
Al₂O₃ MW: 101.96 g/mol Boiling Pt: 2977 °C (760 mmHg) Melting Pt: 2030 °C Density: 3.97 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 REACH: 01-2119529248-35 Merck Index: 13,00355 |
Specification Test Results
Appearance | White Powder |
Sodium oxide (Na2O) | ≤0.1 % |
Silicon dioxide (SiO2) | ≤0.04 % |
Aluminium oxide (Al2O3) | ≥99.70 % |
Iron(III)oxide (Fe2O3) | ≤0.03 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...