Calcium carbonate 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Marble

AC43719-0050 AC43719-0250
TS43719-0050EA 59 USD
TS43719-0050 TS43719-0250
Calcium carbonate 99.999% (trace metals basis)
Calcium carbonate
Formula: CaCO₃
Melting Pt: 825 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric 39 to 42 %
Total trace metal impurities ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR