Calcium carbonate 98+%, heavy powder, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Marble

AC12467-0010 AC12467-0025 AC12467-0100
TS12467-0010EA 83 USD
TS12467-0010 TS12467-0025 TS12467-0100
Calcium carbonate 98+%, heavy powder, pure
Calcium carbonate
Formula: CaCO₃
MW: 100.09 g/mol
Melting Pt: 825 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now

Specification Test Results

Appearance White to off-white Powder
Titration Complexometric ≥98.0 %
Titration with HClO4 0.40 to 0.60 M
NMR ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR