Copper(II) chloride dihydrate 98%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Copper dichloride dihydrate, Cupric chloride dihydrate

AC20634-0025 AC20634-5000
TS20634-0025EA 270 USD
TS20634-0025 TS20634-5000
Copper(II) chloride dihydrate 98%, pure
Copper (II) chloride dihydrate
Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Melting Pt: 100 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
EINECS: 231-210-2
UN: 2802
ADR: 8 III
Merck Index: 13,02660

Order Now

Specification Test Results

Appearance Blue-green to blue Crystalline powder or needles
Titration Complexometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR