Palladium 5% on calcium carbonate lead poisoned (3.5% Pb)

Supplier: Thermo Scientific Chemicals
Danger

AC19507-0100 AC19507-0500
TS19507-0100EA 165 USD
TS19507-0100 TS19507-0500
Palladium 5% on calcium carbonate lead poisoned (3.5% Pb)
Palladium
Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Gray to Gray-black Powder
Assay 5 % (Pd)
Water ≤5.0 %
Additional info poisoned with 3.5% lead


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR