Palladium 5% on alumina, dry, powder for use as a catalyst

Supplier: Thermo Scientific Chemicals

AC29303-0500 AC29303-0100
TS29303-0500EA 795 USD
TS29303-0500 TS29303-0100
Palladium 5% on alumina, dry, powder for use as a catalyst
Palladium
Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now

Specification Test Results

Appearance Silver gray to black Powder
Assay 4.0 to 6.0 % (Pd)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR