Zinc oxide 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zinci oxidumZinc (II) oxide

AC31579-0250 AC31579-1000
TS31579-0250EA 116 USD
TS31579-0250 TS31579-1000
Zinc oxide 99.999% (trace metals basis)
Zinc oxide
Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.6 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200

Order Now

Specification Test Results

Appearance White to light yellow Powder and/or granules
Total trace metal impurities ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR