Sodium percarbonate

Supplier: Thermo Scientific Chemicals
Danger

AC37073-0025 AC37073-0100 AC37073-5000
TS37073-0025EA 116 USD
TS37073-0025 TS37073-0100 TS37073-5000
Sodium percarbonate
Sodium percarbonate
Formula: 2Na₂CO₃·3H₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00043204
CAS Number: 15630-89-4
UN: 3378
ADR: 5.1,III

Order Now

Specification Test Results

Appearance White Granular powder or granules
Titration with KMnO4 12.5 to 14.5 % (Available Oxygen)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR