Phosphonitrilic chloride trimer 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cyclic Phosphonitrilic Chloride Trimer, Hexachlorocyclotriphosphazene

AC17428-0050 AC17428-0250 AC17428-1000
TS17428-0050EA 21 USD
TS17428-0050 TS17428-0250 TS17428-1000
Phosphonitrilic chloride trimer 98%
Phosphonitrilic chloride trimer
Formula: Cl₆N₃P₃
Melting Pt: 111…115 °C
Density: 1.98 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00006474
CAS Number: 940-71-6
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 111 to 115 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR