Zirconium(IV) ethoxide 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC33545-0050
AC33545-0250
TS33545-0050EA
69
USD
TS33545-0050
TS33545-0250
Zirconium(IV) ethoxide 99%
Zirconium(IV) ethoxide
Formula:
C₈H₂₀O₄Zr Boiling Pt: 235 °C (7mbar) Melting Pt: 200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015654 CAS Number: 18267-08-8 UN: 3181 ADR: 4.1,II |
Specification Test Results
Appearance | White Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Assay | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...