3-(Trimethoxysilyl)propyl methacrylate 98%
Supplier: Thermo Scientific Chemicals
AC21655-0050
AC21655-1000
AC21655-5000
TS21655-0050EA
45
USD
TS21655-0050
TS21655-1000
TS21655-5000
3-(Trimethoxysilyl)propyl methacrylate 98%
3-(Trimethoxysilyl)propyl methacrylate
Formula:
C₁₀H₂₀O₅Si Boiling Pt: 253 °C (1017hPa) Density: 1.04 g/cm³ Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008593 CAS Number: 2530-85-0 EINECS: 219-785-8 |
Specification Test Results
Appearance | Clear liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.4300 to 1.4320 (20°C, 589 nm) |
GC | ≥97.5 % |
Color scale | ≤70 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...