Mercury (II) oxide, yellow 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC20573-1000
AC20573-5000
TS20573-1000EA
102
USD
TS20573-1000
TS20573-5000
Mercury (II) oxide, yellow 99+%
Mercury (II) oxide
Formula:
HgO Melting Pt: 500 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011045 CAS Number: 21908-53-2 EINECS: 244-654-7 UN: 1641 ADR: 6.1,II Merck Index: 13,05908 |
Specification Test Results
Appearance | Yellow to orange Powder |
Titration Complexometric | ≥99.0 % |
Residue after ignition | ≤0.05 % (substances not reducible with HCOOH) |
Iron (Fe) | ≤50 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...