Aminoguanidine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aminoguanidine HCl, Aminoguanidine Hydrochloride, Guanylhydrazine hydrochloride

AC36891-0250 AC36891-1000
TS36891-0250EA 58 USD
TS36891-0250 TS36891-1000
Aminoguanidine hydrochloride 98%
Aminoguanidine hydrochloride
Formula: CH₇ClN₄
Melting Pt: >160 °C
Storage Temperature: Ambient
MDL Number: MFCD00039074
CAS Number: 1937-19-5
EINECS: 217-707-7

Order Now

Specification Test Results

Appearance White to off-white Crystals
Infrared spectrum Conforms
Melting point ≥160 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR