Vanadyl(IV) acetylacetonate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Vanadium(IV) oxide acetylacetonate, Oxobis(2,4-pentanedionato-O,O')vanadium (IV), Vanadium(IV) bis(acetylacetonato)oxide, Vanadyl acetylacetonate, Di(2,4-pentandionato-O,O')vanadium(IV)oxid, Vanadium (IV) oxide bis(2,4-pentanedionate)

AC19959-0010 AC19959-0500 AC19959-2500
TS19959-0010EA 747 USD
TS19959-0010 TS19959-0500 TS19959-2500
Vanadyl(IV) acetylacetonate 99%
Vanadyl (IV) acetylacetonate
Formula: C₁₀H₁₄O₅V
MW: 265.16 g/mol
Melting Pt: 256…259 °C
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000032
CAS Number: 3153-26-2
EINECS: 221-590-8
UN: 3285
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Green to blue-green Crystalline powder
Infrared spectrum Conforms
Assay ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR