Sodium ethoxide 96%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium ethylate, Sodium ethanolate

AC16859-0025 AC16859-2500 AC16859-5000
TS16859-0025EA 315 USD
TS16859-0025 TS16859-2500 TS16859-5000
Sodium ethoxide 96%, pure
Sodium ethoxide
Formula: C₂H₅NaO
MW: 68.05 g/mol
Melting Pt: 260 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3095
ADR: 8 I

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Assay ≥95 % (NaOC2H5)
Titration with HCl ≥98 % (total NaOC2H5)
NaOH + Na2CO3 (as NaOH) ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR