Tetrabutylammonium tetrahydroborate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetra-n-butylammonium borohydrideTetrabutylammonium borohydrideTetrabutylammonium tetrahydridoborate

AC21290-0500 AC21290-0100
TS21290-0100EA 81.2 USD
TS21290-0100 TS21290-0500
Tetrabutylammonium tetrahydroborate 98%
Tetrabutylammonium tetrahydroborate
Formula: C₁₆H₄₀BN
MW: 257.31 g/mol
Melting Pt: 126…131 °C
MDL Number: MFCD00012035
CAS Number: 33725-74-5
EINECS: 251-658-2
UN: 3131
ADR: 4.3,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR