Ethyl chloroformate 99%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloroformic acid ethyl ester

AC43079-1000 AC43079-8000
TS43079-1000EA 40 USD
TS43079-1000 TS43079-8000
Ethyl chloroformate 99%, AcroSeal™
Ethyl chloroformate
Formula: C₃H₅ClO₂
Boiling Pt: 94 °C (760 mmHg)
Melting Pt: –81 °C
Density: 1.135 g/cm³
Flash Pt: 10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000644
CAS Number: 541-41-3
UN: 1182
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3940 to 1.3960 (20°C, 589 nm)
GC ≤1.0 % Diethyl carbonate
Color scale ≤50 APHA
Free acid (HCl) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR