Ethyl chloroformate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloroformic acid ethyl ester

AC18108-0025 AC18108-0050 AC18108-5000
TS18108-0025EA 391 USD
TS18108-0025 TS18108-0050 TS18108-5000
Ethyl chloroformate 97%
Ethyl chloroformate
Formula: C₃H₅ClO₂
Boiling Pt: 94 °C (760 mmHg)
Melting Pt: –81 °C
Density: 1.135 g/cm³
Flash Pt: 10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000644
CAS Number: 541-41-3
EINECS: 208-778-5
UN: 1182
ADR: 6.1,
Merck Index: 12,03831

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3940 to 1.3960 (20°C, 589 nm)
GC ≥96.0 %
Color scale ≤100 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR