Silver hexafluoroantimonate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Silver hexafluoroantimonate(V)

AC31250-0050 AC31250-0250
TS31250-0050EA 73 USD
TS31250-0050 TS31250-0250
Silver hexafluoroantimonate 98%
Silver hexafluoroantimonate
Formula: AgF₆Sb
Storage Temperature: Ambient
MDL Number: MFCD00003401
CAS Number: 26042-64-8
EINECS: 247-429-1
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Titration Argentometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR