Copper(II) acetylacetonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate
AC11065-0250
AC11065-1000
AC11065-5000
TS11065-0250EA
44
USD
TS11065-0250
TS11065-1000
TS11065-5000
Copper(II) acetylacetonate 98%
Copper (II) acetylacetonate
Formula:
C₁₀H₁₄CuO₄ Boiling Pt: 160 °C (9.8 mmHg) Melting Pt: 245 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00000016 CAS Number: 13395-16-9 EINECS: 236-477-9 |
Specification Test Results
Appearance | Blue to blue-gray Crystalline powder |
Infrared spectrum | Conforms |
Titration with Na2S2O3 | 97.5 to 102.5 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...