Silver tetrafluoroborate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Argentous tetrafluoroborate
AC19709-0010
AC19709-0100
AC19709-0500
TS19709-0010EA
35
USD
TS19709-0010
TS19709-0100
TS19709-0500
Silver tetrafluoroborate 99%
Silver tetrafluoroborate
Formula:
AgBF₄ Melting Pt: 200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003408 CAS Number: 14104-20-2 EINECS: 237-956-5 UN: 3260 ADR: 8,II |
Specification Test Results
Appearance | Beige Crystalline powder |
Assay | ≥43.92 % (BF4) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...