Tantalum(V) oxide 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Ditantalum pentaoxide

AC19417-0100 AC19417-0500
TS19417-0100EA 69 USD
TS19417-0100 TS19417-0500
Tantalum(V) oxide 99.99% (trace metals basis)
Tantalum (V) oxide
Formula: O₅Ta₂
Melting Pt: 1872 °C
MDL Number: MFCD00011254
CAS Number: 1314-61-0
EINECS: 215-238-2
Merck Index: 14,09056

Order Now

Specification Test Results

Appearance White to light beige Powder
Total trace metal impurities ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR