Diyttrium trioxide 99.999% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Yttrium oxide, Yttrium (III) oxide

AC19448-0020 AC19448-0100 AC19448-0500
TS19448-0020EA 29 USD
TS19448-0020 TS19448-0100 TS19448-0500
Diyttrium trioxide 99.999% (trace metals basis)
Diyttrium trioxide
Formula: Y₂O₃
Melting Pt: 2410 °C
Storage Temperature: Ambient
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5

Order Now

Specification Test Results

Appearance White Powder
Assay 99.999 % (Y2O3) On TREO base


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR