Dibutyltin dichloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dichlorodibutylstannane, Di-n-butyltin dichloride, DBTC

AC19487-0250 AC19487-2500
TS19487-0250EA 29 USD
TS19487-0250 TS19487-2500
Dibutyltin dichloride 97%
Dibutyltin dichloride
Formula: C₈H₁₈Cl₂Sn
MW: 303.85 g/mol
Boiling Pt: 275
Melting Pt: 39…41 °C
Density: 1.4 g/cm³
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000518
CAS Number: 683-18-1
EINECS: 211-670-0
UN: 2928
ADR: 6.1 II

Order Now

Specification Test Results

Appearance White to beige Low melting crystalline mass
Infrared spectrum Conforms
Assay ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR