Triethylsilyl trifluoromethanesulfonate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Triethylsilyl trifluoromethanesulfonate, Triethylsilyl trifluoromethanesulphonate

AC31270-0100 AC31270-0500
TS31270-0100EA 109 USD
TS31270-0100 TS31270-0500
Triethylsilyl trifluoromethanesulfonate 99%
Triethylsilyl trifluoromethanesulphonate
Formula: C₇H₁₅F₃O₃SSi
MW: 264.34 g/mol
Boiling Pt: 85…86 °C (12 mmHg)
Density: 1.16 g/cm³
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00000407
CAS Number: 79271-56-0
EINECS: 279-124-4
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.3890 to 1.3920 (20°C, 589 nm)
Titration excess NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR