Potassium formate 96% pract.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Formic acid potassium salt

AC41822-0050 AC41822-1000 AC41822-5000
TS41822-0050EA 385 USD
TS41822-0050 TS41822-1000 TS41822-5000
Potassium formate 96% pract.
Potassium formate
Formula: CHKO₂
MW: 84.12 g/mol
Melting Pt: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00013100
CAS Number: 590-29-4
EINECS: 209-677-9
Merck Index: 12,07795

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 164 to 168 °C
Titration with HClO4 ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR