Terbium trinitrate hexahydrate 99.99+% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Terbium trinitrate 6-hydrate, Terbium(III) nitrate hexahydrate, Terbium(III) nitrate hexahydrate (1:3:6), Terbium (III) nitrate 6-hydrate, Terbium (III) nitrate hexahydrate

AC29579-0020 AC29579-0100
TS29579-0020EA 70 USD
TS29579-0020 TS29579-0100
Terbium trinitrate hexahydrate 99.99+% (trace metals basis)
Terbium trinitrate hexahydrate
Formula: N₃O₉Tb·6H₂O
MW: 453.03
Melting Pt: 89.3
Storage Temperature: Ambient
MDL Number: MFCD00149868
CAS Number: 13451-19-9
EINECS: 233-138-7
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance White Granular powder
Assay ≥99.99 % (TREO base)
Total conc. of imp.as REO ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR