Tin(IV) oxide 99.9% (trace metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide

AC22368-1000 AC22368-5000
TS22368-1000EA 70 USD
TS22368-1000 TS22368-5000
Tin(IV) oxide 99.9% (trace metals basis)
Tin (IV) oxide
Formula: O₂Sn
MW: 150.71 g/mol
Boiling Pt: ∼2500 °C (1013 hPa)
Melting Pt: 1624.85
Density: 6.95 g/cm³
MDL Number: MFCD00011244
CAS Number: 18282-10-5
EINECS: 242-159-0
Merck Index: 14,08778

Order Now

Specification Test Results

Appearance White to light gray Powder
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR