Vanadium(V) oxide 99.6+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide

AC20642-0010 AC20642-2500
TS20642-0010EA 251 USD
TS20642-0010 TS20642-2500
Vanadium(V) oxide 99.6+%
Vanadium (V) oxide
Formula: O₅V₂
MW: 181.88 g/mol
Boiling Pt: 1750
Melting Pt: 690
Density: 3.35 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011457
CAS Number: 1314-62-1
EINECS: 215-239-8
UN: 2862
ADR: 6.1 III
Merck Index: 13,09987

Order Now

Specification Test Results

Appearance Yellow to orange-brown Powder
Assay ≥99.60 %
Chromium (Cr) ≤100 ppm
Iron (Fe) ≤300 ppm
Potassium (K) ≤200 ppm
Silicon (Si) ≤200 ppm
Sodium (Na) ≤200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR