Tetramethylsilane 99.9+% for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TMS

AC13847-0250 AC13847-1000
TS13847-1000EA 159 USD
TS13847-1000 TS13847-0250
Tetramethylsilane 99.9+% for NMR spectroscopy
Tetramethylsilane
Formula: C₄H₁₂Si
MW: 88.22 g/mol
Boiling Pt: 26…28 °C (760 mmHg)
Melting Pt: –99 °C
Density: 0.64 g/cm³
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008274
CAS Number: 75-76-3
EINECS: 200-899-1
UN: 2749
ADR: 3,I

Order Now

Specification Test Results

Appearance Clear colorless Liquid
GC ≥99.90 %
Suitability (for 1H-NMR position ref.) Passes test
Elemental Analysis 26.08 to 28.25 % C
Elemental Analysis 6.09 to 6.59 % N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR