Mercury(II) iodide, red ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC19350-0250
AC19350-1000
AC19350-5000
TS19350-0250EA
66
USD
TS19350-0250
TS19350-1000
TS19350-5000
Mercury(II) iodide, red ACS
Mercury (II) iodide
Formula:
HgI₂ Boiling Pt: 354 °C (760 mmHg) Melting Pt: 259 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011044 CAS Number: 7774-29-0 UN: 1638 ADR: 6.1,II |
Specification Test Results
Appearance | Red Crystalline powder |
Titration Iodometric | ≥99.0 % (Dried basis) |
Solubility | (In potassium iodide soln.) Passes test |
Mercurous Mercury (as Hg) | ≤0.1 % |
Soluble Hg salts (as Hg) | ≤0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...