Platinum 10% on activated carbon wetted with ca. 50% water

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

AC19524-0010 AC19524-0100 AC19524-0500
TS19524-0010EA 40 USD
TS19524-0010 TS19524-0100 TS19524-0500
Platinum 10% on activated carbon wetted with ca. 50% water
Platinum
Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1

Order Now

Specification Test Results

Appearance Black Powder
Moisture 45 % to 60 %
Platinum (Pt) 9.8 to 10.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR