Triethyl borate 97%

Supplier: Thermo Scientific Chemicals
Danger

AC13958-0010 AC13958-2500
TS13958-0010EA 339 USD
TS13958-0010 TS13958-2500
Triethyl borate 97%
Triethyl borate
Formula: C₆H₁₅BO₃
MW: 145.99 g/mol
Boiling Pt: 117
Melting Pt: -84.8
Density: 0.858 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00012846
CAS Number: 150-46-9
EINECS: 205-760-9
UN: 1176
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3705 to 1.3775 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR