Niobium(V) oxide 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Diniobium pentaoxide, Niobium pentoxide

AC19363-0100 AC19363-0500 AC19363-2500
TS19363-0100EA 42 USD
TS19363-0100 TS19363-0500 TS19363-2500
Niobium(V) oxide 99.99% (trace metals basis)
Niobium (V) oxide
Formula: Nb₂O₅
Melting Pt: 1520 °C
Density: 4.47 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011128
CAS Number: 1313-96-8
EINECS: 215-213-6
Merck Index: 14,06559

Order Now

Specification Test Results

Appearance White to off-white Powder
Total trace metal impurities ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR