Nickel(II) chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel dichloride

AC38732-0500 AC38732-2500
TS38732-0500EA 58 USD
TS38732-0500 TS38732-2500
Nickel(II) chloride 98%
Nickel (II) chloride
Formula: Cl₂Ni
Melting Pt: 1001 °C
Storage Temperature: Ambient
MDL Number: MFCD00011142
CAS Number: 7718-54-9
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light orange to brown Powder
Titration Argentometric ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR