Neodymium(III) oxide 99.9% (trace metals basis)
Supplier: Thermo Scientific Chemicals
AC21158-0250
AC21158-1000
TS21158-0250EA
74
USD
TS21158-0250
TS21158-1000
Neodymium(III) oxide 99.9% (trace metals basis)
Neodymium (III) oxide
Formula:
Nd₂O₃ Boiling Pt: 3760 °C (1013 hPa) Melting Pt: 2270 °C Density: 7.24 g/cm³ |
MDL Number:
MFCD00011134 CAS Number: 1313-97-9 EINECS: 215-214-1 Merck Index: 14,06450 |
Specification Test Results
Appearance | Blue Powder |
Total trace metal impurities | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...