Sodium formate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Formic acid sodium salt

AC14823-0010 AC14823-0051
TS14823-0010EA 50.92 USD
TS14823-0010 TS14823-0051
Sodium formate 99%
Sodium formate
Formula: HCO₂Na
MW: 68.01 g/mol
Boiling Pt: 360 °C (760 mmHg)
Melting Pt: 258…261 °C
Storage Temperature: Ambient
MDL Number: MFCD00013101
CAS Number: 141-53-7
EINECS: 205-488-0
REACH: 01-2119486468-21
Merck Index: 12,08765

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 253 to 261 °C
Water ≤0.5 %
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR