Potassium methanolate 0.1 N in toluene/methanol, AcroSeal™, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC42910-1000
TS42910-1000EA
50
USD
TS42910-1000
Potassium methanolate 0.1 N in toluene/methanol, AcroSeal™, pure
Potassium methanolate
Formula:
CH₃KO MW: 70.13 g/mol Density: 0.85 g/cm³ Flash Pt: 7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012416 CAS Number: 865-33-8 UN: 1992 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Solution |
Strength | 0.095 to 0.105 N |
Titration Complexometric | 23.5 to 26.0 % (Zinc) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...