Potassium methanolate 0.1 N in toluene/methanol, pure

Supplier: Thermo Scientific Chemicals
Danger

AC24060-5000
TS24060-5000EA 231 USD
TS24060-5000
Potassium methanolate 0.1 N in toluene/methanol, pure
Potassium methanolate
Formula: CH₃KO
MW: 70.13 g/mol
Density: 0.85 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00012416
CAS Number: 865-33-8
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration with HCl 0.095 to 0.105 N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR