Manganese(IV) oxide 88%, electrolytically precipitated, active
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid
AC20319-0025
AC20319-1000
AC20319-5000
TS20319-0025EA
334
USD
TS20319-0025
TS20319-1000
TS20319-5000
Manganese(IV) oxide 88%, electrolytically precipitated, active
Manganese (IV) oxide
Formula:
MnO₂ Melting Pt: 535 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003463 CAS Number: 1313-13-9 EINECS: 215-202-6 UN: 1479 ADR: 5.1,II Merck Index: 14,05730 |
Specification Test Results
Appearance | Black Powder |
Titration Iodometric | ≥83 % (on dried substance) |
Additional info | Oxidation activity: >60% (yield) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...