Magnesium sulfate heptahydrate 99.5% for analysis

Supplier: Thermo Scientific Chemicals

Synonyms: sulphuric acid magnesium salt heptahydrate, sulfuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

AC21311-0025 AC21311-0250 AC21311-5000
TS21311-0025EA 314 USD
TS21311-0025 TS21311-0250 TS21311-5000
Magnesium sulfate heptahydrate 99.5% for analysis
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 1124
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Order Now

Specification Test Results

Appearance White Crystals
Titration Complexometric ≥99.4 %
pH 5 to 8 (5 % at 25°C)
Chloride (Cl) ≤5 ppm
Cadmium (Cd) ≤5 ppm
Calcium (Ca) ≤50 ppm
Cobalt (Co) ≤5 ppm
Iron (Fe) ≤5 ppm
Manganese (Mn) ≤5 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤200 ppm
Strontium (Sr) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR