Magnesium sulfate heptahydrate 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals

Synonyms: sulfuric acid magnesium salt heptahydrateBitter saltsulphuric acid magnesium salt heptahydrateEpsom salt

AC12490-0025 AC12490-0100 AC12490-0010
TS12490-0025EA 180 USD
TS12490-0025 TS12490-0100 TS12490-0010
Magnesium sulfate heptahydrate 99+%, Extra Pure
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Melting Pt: 770 °C
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Complexometric ≥99.0 % (on dry substance)
Loss on drying 48.0 % to 52.0 % (450°C)
pH 5.0 to 9.2 (5 % at 20°C)
Chloride (Cl) ≤200 ppm
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR