Copper(II) oxide, powder ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Copper monoxide granulatedCopper oxideCupric oxide

AC40586-0250 AC40586-2500 AC40586-0010
TS40586-0250EA 28.7 USD
TS40586-0250 TS40586-0010 TS40586-2500
Copper(II) oxide, powder ACS
Copper (II) oxide
Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 1326 °C
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9,III
Merck Index: 13,02674

Order Now

Specification Test Results

Appearance Black to brown-black Powder
Titration Complexometric ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤200 ppm
Calcium (Ca) ≤100 ppm
Iron (Fe) ≤500 ppm
Potassium (K) ≤200 ppm
Sodium (Na) ≤500 ppm
Carbon compounds (as C) ≤0.01 %
Insoluble matter ≤0.02 % (in dilute HCl)
Nitrogen compounds (as N) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR